Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526999_s1
          See other OLFR702 GE Assays ›
          Gene Symbol:
          Olfr702
          Gene Name:
          olfactory receptor 702
          Gene Aliases:
          MOR260-4
          Chromosome Location:
          Chr.7: 106823489 - 106826743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526999_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr702
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146597.2 NP_666808.1 2 561 120
          GenBank mRNA BC120573.1 - 1 341 40129496 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004021.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr702

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957792 NM_146597.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957792 BC120573.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700310 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111541 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957792 NM_146597.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957792 BC120573.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P323701 NM_146597.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P323701 BC120573.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396785 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111541 - -
          Whole Mouse Genome G4122A A_51_P323701 NM_146597.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P323701 BC120573.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140821 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06526552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06526536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06473628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06473634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06473629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06086123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06230332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566605 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566603 - -
          Affymetrix Mouse Clariom S Array PSR0700033451.mm.1 NM_146597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033451.mm.1 NM_146597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033451.mm.1 TC0700004021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033452.mm.1 NM_146597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033452.mm.1 NM_146597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033452.mm.1 TC0700004021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033453.mm.1 NM_146597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033453.mm.1 NM_146597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033453.mm.1 TC0700004021.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 377524 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740070 - -
          Illumina mouse-6 v1.0 expression beadchip 1740070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252706 - -
          mouse-6 v1.1 (Illumina) scl8846.1.1_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8846.1.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252706 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0