Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527056_s1
          See other OLFR716 GE Assays ›
          Gene Symbol:
          Olfr716
          Gene Name:
          olfactory receptor 716
          Gene Aliases:
          MOR260-2
          Chromosome Location:
          Chr.7: 107147318 - 107148262 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527056_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr716
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146604.1 NP_666815.1 1 856 113
          GenBank mRNA BC120510.1 - 1 948 40129433 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001572.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr716

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211978 NM_146604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211978 NM_146604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121816 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211978 NM_146604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211978 BC120510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211978 NM_146604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211978 NM_146604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211978 NM_146604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121816 - -
          Whole Mouse Genome G4122A A_51_P211978 NM_146604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211978 BC120510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211978 NM_146604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211978 BC120510.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108461 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25398068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06126788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06652686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06028622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06652687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25398076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556034 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556033 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556041 - -
          Affymetrix Mouse Clariom S Array PSR0700013164.mm.1 TC0700001572.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013165.mm.1 TC0700001572.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013165.mm.1 NM_146604.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013165.mm.1 NM_146604.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 649849 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3940463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222834 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222834 - -
          mouse-6 v1.1 (Illumina) scl32228.1.1_213-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32228.1.1_213-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222834 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222834 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32228.1.1_213-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0