Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527105_s1
          See other OLFR1443 GE Assays ›
          Gene Symbol:
          Olfr1443
          Gene Name:
          olfactory receptor 1443
          Gene Aliases:
          MOR202-8
          Chromosome Location:
          Chr.19: 12678166 - 12683851 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527105_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1443
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146698.2 NP_666909.1 3 1146 97
          GenBank mRNA BC065104.1 - 3 1146 6841907 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1443

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996711 NM_146698.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996711 BC065104.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776626 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105051 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996711 NM_146698.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996711 BC065104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262131 NM_146698.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262131 BC065104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P436662 NM_146698.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P436662 BC065104.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09109039 - -
          Whole Mouse Genome G4122A A_52_P436662 NM_146698.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P436662 BC065104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262131 NM_146698.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262131 BC065104.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118379 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14259674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14259664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14259669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14259665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14259672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14330027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14259672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14308298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14453105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14232640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461753 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461750 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440685 - -
          Illumina mouse-6 v1.0 expression beadchip 3440685 - -
          Illumina mouse-6 v1.0 expression beadchip 3870600 - -
          Illumina mouse-6 v1.0 expression beadchip 4560026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230566 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2597700 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2690703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230566 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2690703 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2597700 - -
          mouse-6 v1.1 (Illumina) scl0258693.3_79-S - -
          mouse-6 v1.1 (Illumina) scl00258693.2_13-S - -
          mouse-6 v1.1 (Illumina) scl53338.3.30_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440685 - -
          Illumina Sentrix mouse-6.v1 bead array 3870600 - -
          Illumina Sentrix mouse-6.v1 bead array 4560026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53338.3.30_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258693.2_13-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258693.3_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440685 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2891065 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230566 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2597700 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2690703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2690703 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2891065 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2597700 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230566 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53338.3.30_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline