Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527139_s1
          See other OLFR1444 GE Assays ›
          Gene Symbol:
          Olfr1444
          Gene Name:
          olfactory receptor 1444
          Gene Aliases:
          MOR202-4
          Chromosome Location:
          Chr.19: 12857421 - 12862765 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527139_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1444
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146702.1 NP_666913.1 1 254 67
          GenBank mRNA BC119059.1 - 1 289 8733808 67
          BC125619.1 - 1 289 40130134 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000257.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1444

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157165 NM_146702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157165 BC119059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157165 BC125619.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487777 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120466 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157165 NM_146702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157165 BC119059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157165 BC125619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425010 NM_146702.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425010 BC119059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425010 BC125619.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120466 - -
          Whole Mouse Genome G4122A A_51_P425010 NM_146702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425010 BC119059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425010 BC125619.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122665 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14170850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14170852 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14170843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14170840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14365850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14507900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14145002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14300880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14170852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14160062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461779 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461777 - -
          Affymetrix Mouse Clariom S Array PSR1900003015.mm.1 NM_146702.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003015.mm.1 NM_146702.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003015.mm.1 TC1900000257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003014.mm.1 TC1900000257.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 467119 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 70292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648542 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648542 - -
          mouse-6 v1.1 (Illumina) scl6295.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 70292 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6295.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2648542 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline