Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527532_s1
          See other OLFR661 GE Assays ›
          Gene Symbol:
          Olfr661
          Gene Name:
          olfactory receptor 661
          Gene Aliases:
          MOR40-4
          Chromosome Location:
          Chr.7: 104688017 - 104688976 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527532_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr661
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146748.1 NP_666959.1 1 849 116
          GenBank mRNA BC128005.1 - 1 853 40135960 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr661

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519632 NM_146748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519632 NM_146748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855301 AK016459.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855301 BC061039.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855301 NM_026624.2 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135019 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519632 NM_146748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519632 AF121983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519632 NM_146748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P519632 NM_146748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P519632 NM_146748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135019 - -
          Whole Mouse Genome G4122A A_51_P519632 NM_146748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519632 AF121983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519632 NM_146748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519632 AF121983.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105235 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06153126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06281538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06091883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06153124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06091895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06281555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06281541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05973963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06091894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06281536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555888 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555887 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555964 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688387 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688387 - -
          mouse-6 v1.1 (Illumina) scl32251.1.399_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32251.1.399_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2688387 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2688387 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0