Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527580_s1
          See other OLFR556 GE Assays ›
          Gene Symbol:
          Olfr556
          Gene Name:
          olfactory receptor 556
          Gene Aliases:
          MOR41-1
          Chromosome Location:
          Chr.7: 102669901 - 102670923 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527580_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr556
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146754.2 NP_666965.2 1 64 83
          GenBank mRNA BC138388.1 - 1 64 8861408 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001464.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr556

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134456 NM_146754.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134456 BC138388.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135552 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134456 NM_146754.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134456 BC138388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517662 NM_146754.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517662 BC138388.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379953 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135552 - -
          Whole Mouse Genome G4122A A_51_P517662 NM_146754.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517662 BC138388.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102906 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06575637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06004710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06453902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06329874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06106302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06261345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555733 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555731 - -
          Affymetrix Mouse Clariom S Array PSR0700012689.mm.1 TC0700001464.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012689.mm.1 NM_146754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012689.mm.1 NM_146754.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 391942 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 610091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2690884 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2690884 - -
          mouse-6 v1.1 (Illumina) scl9411.1.1_287-S - -
          Illumina Sentrix mouse-6.v1 bead array 610091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9411.1.1_287-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2690884 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2690884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline