Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527629_s1
          See other OLFR214 GE Assays ›
          Gene Symbol:
          Olfr214
          Gene Name:
          olfactory receptor 214
          Gene Aliases:
          MOR119-1
          Chromosome Location:
          Chr.6: 116556427 - 116557389 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527629_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr214
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146759.1 NP_666970.1 1 62 62
          GenBank mRNA BC138076.1 - 1 84 8861095 62
          BC138077.1 - 1 84 8861096 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001306.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr214

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174756 BC138076.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174756 BC138077.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174756 NM_146759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174756 BC138076.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174756 BC138077.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174756 NM_146759.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P308625 BC138076.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P308625 BC138077.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P308625 NM_146759.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777132 - -
          Whole Mouse Genome G4122A A_51_P308625 BC138076.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308625 BC138077.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P308625 NM_146759.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130776 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04998711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05155805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05155809 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04998712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05185385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05185386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05336727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05805539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P04998718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05336732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05261163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777155 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541070 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541069 - -
          Affymetrix Mouse Clariom S Array PSR0600010369.mm.1 TC0600001306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600010369.mm.1 NM_146759.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600010369.mm.1 NM_146759.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817409 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220184 - -
          Illumina mouse-6 v1.0 expression beadchip 5220184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220184 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659858 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659858 - -
          mouse-6 v1.1 (Illumina) scl29591.1.1_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220184 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29591.1.1_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659858 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2659858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline