Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527749_s1
          See other OLFR508 GE Assays ›
          Gene Symbol:
          Olfr508
          Gene Name:
          olfactory receptor 508
          Gene Aliases:
          MOR204-6
          Chromosome Location:
          Chr.7: 108629994 - 108630926 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527749_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr508
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146773.1 NP_666984.1 1 50 106
          GenBank mRNA BC130241.1 - 1 54 40142176 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001602.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr508

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079806 NM_146773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079806 BC130241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079806 NM_146773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079806 BC130241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146593 NM_146773.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P146593 BC130241.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404053 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123981 - -
          Whole Mouse Genome G4122A A_51_P146593 NM_146773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P146593 BC130241.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108104 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123981 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05922145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05922147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06348985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05922159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06348987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05922148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06348984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25403999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06129833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06472228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05985426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06000126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556159 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556158 - -
          Affymetrix Mouse Clariom S Array PSR0700013287.mm.1 TC0700001602.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013287.mm.1 NM_146773.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013287.mm.1 NM_146773.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 637453 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2190079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250519 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250519 - -
          mouse-6 v1.1 (Illumina) scl32213.1.139_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32213.1.139_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250519 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline