Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527823_s1
          See other OLFR922 GE Assays ›
          Gene Symbol:
          Olfr922
          Gene Name:
          olfactory receptor 922
          Gene Aliases:
          MOR161-3
          Chromosome Location:
          Chr.9: 38815408 - 38816539 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527823_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr922
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146781.2 NP_666992.1 1 563 84
          GenBank mRNA BC119109.1 - 1 563 8733858 84
          BC125623.1 - 1 563 40130138 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000417.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr922

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061954 BC125623.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061954 NM_146781.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061954 BC119109.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157427 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061954 BC125623.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061954 NM_146781.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061954 BC119109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P222725 BC125623.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P222725 NM_146781.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P222725 BC119109.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436184 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157427 - -
          Whole Mouse Genome G4122A A_51_P222725 BC125623.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P222725 NM_146781.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P222725 BC119109.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140668 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08067226 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08067218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08067211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08067202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08067207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08067212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08067213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436176 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584465 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584462 - -
          Affymetrix Mouse Clariom S Array PSR0900002865.mm.1 NM_146781.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002865.mm.1 NM_146781.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002865.mm.1 TC0900000417.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 629266 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520390 - -
          Illumina mouse-6 v1.0 expression beadchip 6520390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246083 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246083 - -
          mouse-6 v1.1 (Illumina) scl37084.2.98_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37084.2.98_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246083 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246083 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0