Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527952_s1
          See other OLFR1499 GE Assays ›
          Gene Symbol:
          Olfr1499
          Gene Name:
          olfactory receptor 1499
          Gene Aliases:
          MOR211-2
          Chromosome Location:
          Chr.19: 13814644 - 13815588 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527952_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1499
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146796.1 NP_667007.1 1 431 72
          GenBank mRNA BC132138.1 - 1 501 40130415 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001188.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1499

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170090 BC132138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170090 BC132138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143366 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170090 BC132138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170090 NM_146796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170090 BC132138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170090 BC132138.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170090 BC132138.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965992 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143366 - -
          Whole Mouse Genome G4122A A_51_P170090 BC132138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170090 NM_146796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170090 BC132138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P170090 NM_146796.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143366 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14239693 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14239688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14239691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14239692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14381936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14188373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31966004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14427723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14381923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31966005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14332941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14332952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965945 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466367 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466366 - -
          Affymetrix Mouse Clariom S Array PSR1900010880.mm.1 TC1900001188.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010882.mm.1 TC1900001188.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010881.mm.1 NM_146796.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010881.mm.1 NM_146796.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010881.mm.1 TC1900001188.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 495434 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4280300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215595 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215595 - -
          mouse-6 v1.1 (Illumina) scl6100.1.1_70-S - -
          Illumina Sentrix mouse-6.v1 bead array 4280300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6100.1.1_70-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215595 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215595 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline