Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528092_s1
          See other OLFR665 GE Assays ›
          Gene Symbol:
          Olfr665
          Gene Name:
          olfactory receptor 665
          Gene Aliases:
          MOR34-7
          Chromosome Location:
          Chr.7: 104880709 - 104881659 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528092_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr665
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146814.1 NP_667025.1 1 62 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001536.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr665

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095683 NM_146814.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901385 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160740 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095683 NM_146814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P216990 NM_146814.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160740 - -
          Whole Mouse Genome G4122A A_51_P216990 NM_146814.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134685 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06654893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06355890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06238577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06247726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06477444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06247725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555900 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555899 - -
          Affymetrix Mouse Clariom S Array PSR0700012922.mm.1 TC0700001536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012922.mm.1 NM_146814.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012922.mm.1 NM_146814.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 567978 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2970162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223342 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223342 - -
          mouse-6 v1.1 (Illumina) scl32249.1.1_271-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32249.1.1_271-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223342 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0