Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528182_s1
          See other OLFR1184 GE Assays ›
          Gene Symbol:
          Olfr1184
          Gene Name:
          olfactory receptor 1184
          Gene Aliases:
          MOR225-3
          Chromosome Location:
          Chr.2: 88486734 - 88487669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528182_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1184
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146823.1 NP_667034.1 1 449 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001320.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1184

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167236 NM_146823.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133271 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167236 NM_146823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434427 NM_146823.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21409974 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133271 - -
          Whole Mouse Genome G4122A A_51_P434427 NM_146823.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138957 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139830 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133271 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01153015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01606184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01606174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01931886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21410030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01606186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01406908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02248330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21410000 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473595 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473594 - -
          Affymetrix Mouse Clariom S Array PSR0200010098.mm.1 NM_146823.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010098.mm.1 NM_146823.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010098.mm.1 TC0200001320.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 469754 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7050368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642367 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642367 - -
          mouse-6 v1.1 (Illumina) scl4901.1.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4901.1.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2855467 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2855467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0