Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528191_s1
          See other OLFR273 GE Assays ›
          Gene Symbol:
          Olfr273
          Gene Name:
          olfactory receptor 273
          Gene Aliases:
          MOR262-8
          Chromosome Location:
          Chr.4: 52855558 - 52856511 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528191_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr273
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146824.1 NP_667035.1 1 448 88
          GenBank mRNA BC125463.1 - 1 509 40129978 88
          BC125465.1 - 1 509 40129980 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002662.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr273

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993303 BC125465.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993303 NM_146824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993303 BC125463.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140440 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993303 BC125465.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993303 NM_146824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993303 BC125463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289720 BC125465.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289720 NM_146824.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P289720 BC125463.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22919756 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140440 - -
          Whole Mouse Genome G4122A A_51_P289720 BC125465.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P289720 NM_146824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P289720 BC125463.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03880234 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03941048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03880242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03941051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03941044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03941047 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03880239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03880230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22919733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03880245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03851096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03880237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03531895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03301519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03868270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03941046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512942 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512941 - -
          Affymetrix Mouse Clariom S Array PSR0400021589.mm.1 TC0400002662.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021588.mm.1 TC0400002662.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021588.mm.1 NM_146824.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021588.mm.1 NM_146824.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 877491 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840300 - -
          Illumina mouse-6 v1.0 expression beadchip 6840300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255796 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255796 - -
          mouse-6 v1.1 (Illumina) scl2443.1.1_212-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2443.1.1_212-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255796 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255796 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0