Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528507_s1
          See other OLFR992 GE Assays ›
          Gene Symbol:
          Olfr992
          Gene Name:
          olfactory receptor 992
          Gene Aliases:
          MOR203-1
          Chromosome Location:
          Chr.2: 85399602 - 85400531 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528507_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr992
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146865.1 NP_667076.1 1 145 59
          GenBank mRNA BC115959.1 - 1 144 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003893.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr992

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P398069 NM_146865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P398069 NM_146865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170880 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P398069 NM_146865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P398069 BC115959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P398069 NM_146865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398069 NM_146865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398069 NM_146865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170880 - -
          Whole Mouse Genome G4122A A_51_P398069 NM_146865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398069 BC115959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398069 NM_146865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398069 BC115959.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121121 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134061 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21397685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02100794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01576567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01611379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01576576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01185709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21397735 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484512 - -
          Affymetrix Mouse Clariom S Array PSR0200033683.mm.1 NM_146865.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033683.mm.1 NM_146865.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033683.mm.1 TC0200003893.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 727920 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4590154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240497 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240497 - -
          mouse-6 v1.1 (Illumina) scl4362.1.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4362.1.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2936691 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2936691 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0