Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528585_s1
          See other OLFR895 GE Assays ›
          Gene Symbol:
          Olfr895
          Gene Name:
          olfactory receptor 895
          Gene Aliases:
          MOR170-1
          Chromosome Location:
          Chr.9: 38268001 - 38269753 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528585_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr895
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146875.2 NP_667086.2 1 437 81
          GenBank mRNA BC127996.1 - 1 441 81
          BC152319.1 - 1 441 40135771 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000393.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr895

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968502 BC127996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968502 BC152319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968502 NM_146875.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160098 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968502 BC127996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968502 BC152319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968502 NM_146875.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403504 BC127996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403504 BC152319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P403504 NM_146875.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434191 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160098 - -
          Whole Mouse Genome G4122A A_51_P403504 BC127996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403504 BC152319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403504 NM_146875.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101457 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07921153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07901337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07596969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07866714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08054934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07946383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08060973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07652537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434214 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584389 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584388 - -
          Affymetrix Mouse Clariom S Array PSR0900002789.mm.1 NM_146875.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002789.mm.1 NM_146875.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002789.mm.1 TC0900000393.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 718962 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070438 - -
          Illumina mouse-6 v1.0 expression beadchip 4070438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632950 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632950 - -
          mouse-6 v1.1 (Illumina) scl0258875.1_58-S - -
          Illumina Sentrix mouse-6.v1 bead array 4070438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258875.1_58-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2632950 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632950 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0258875.1_58-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline