Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528632_s1
          See other OLFR876 GE Assays ›
          Gene Symbol:
          Olfr876
          Gene Name:
          olfactory receptor 876
          Gene Aliases:
          MOR161-1
          Chromosome Location:
          Chr.9: 37803889 - 37804936 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528632_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr876
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146883.2 NP_667094.2 1 485 89
          GenBank mRNA BC120726.1 - 1 485 40129649 89
          BC120732.1 - 1 485 40129655 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000375.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr876

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005630 BC120726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005630 BC120732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005630 BC120726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005630 BC120732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P370372 NM_146883.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P370372 BC120726.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P370372 BC120732.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26432484 - -
          Whole Mouse Genome G4122A A_51_P370372 NM_146883.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370372 BC120726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370372 BC120732.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122434 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156579 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156563 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26432482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07797675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08156552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26432468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07797668 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584357 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584356 - -
          Affymetrix Mouse Clariom S Array PSR0900002756.mm.1 NM_146883.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002756.mm.1 NM_146883.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002756.mm.1 TC0900000375.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 586924 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2350373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640485 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640485 - -
          mouse-6 v1.1 (Illumina) scl0258883.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258883.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640485 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0