Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528823_s1
          See other OLFR853 GE Assays ›
          Gene Symbol:
          Olfr853
          Gene Name:
          olfactory receptor 853
          Gene Aliases:
          MOR154-1
          Chromosome Location:
          Chr.9: 19537011 - 19537928 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528823_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr853
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146906.1 NP_667117.1 1 562 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr853

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123057 NM_146906.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123057 NM_146906.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P256294 NM_146906.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26340245 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101311 - -
          Whole Mouse Genome G4122A A_51_P256294 NM_146906.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138471 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08024545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07497941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08176446 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08176442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26340195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07497939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08176440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07520643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26340232 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591197 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591196 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 492257 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236688 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236688 - -
          mouse-6 v1.1 (Illumina) scl6693.1.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6693.1.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ggg7m:80/100.66.79.104:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0