Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528836_s1
          See other OLFR1378 GE Assays ›
          Gene Symbol:
          Olfr1378
          Gene Name:
          olfactory receptor 1378
          Gene Aliases:
          MOR129-2
          Chromosome Location:
          Chr.11: 50968500 - 50969967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528836_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1378
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146910.1 NP_667121.1 1 53 83
          GenBank mRNA BC106990.2 - 1 52 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000553.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1378

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339564 NM_146910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339564 NM_146910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102373 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339564 NM_146910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339564 BC106990.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339564 NM_146910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339564 NM_146910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339564 NM_146910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791589 - -
          Whole Mouse Genome G4122A A_51_P339564 NM_146910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339564 BC106990.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339564 NM_146910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339564 BC106990.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136602 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09550843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09550851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09550860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09550856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09390473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09164029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09164016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09164017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09094664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09137356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09164013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09294308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375788 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375787 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375791 - -
          Affymetrix Mouse Clariom S Array PSR1100004294.mm.1 NM_146910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004294.mm.1 NM_146910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100004294.mm.1 TC1100000553.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 444026 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710725 - -
          Illumina mouse-6 v1.0 expression beadchip 3710725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735402 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735402 - -
          mouse-6 v1.1 (Illumina) scl41604.1.1_221-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41604.1.1_221-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2735408 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline