Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528897_s1
          See other OLFR1179 GE Assays ›
          Gene Symbol:
          Olfr1179
          Gene Name:
          olfactory receptor 1179
          Gene Aliases:
          MOR225-2
          Chromosome Location:
          Chr.2: 88402009 - 88402932 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528897_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1179
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146917.2 NP_667128.2 1 442 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004018.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1179

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084480 NM_146917.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796774 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160369 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084480 NM_146917.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265822 NM_146917.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160369 - -
          Whole Mouse Genome G4122A A_51_P265822 NM_146917.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132129 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21409574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02099428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01438748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02037853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01340929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02099429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01651341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01481075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21409633 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484740 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484739 - -
          Affymetrix Mouse Clariom S Array PSR0200033949.mm.1 NM_146917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033949.mm.1 NM_146917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033949.mm.1 TC0200004018.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4610131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004610131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769462 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769462 - -
          mouse-6 v1.1 (Illumina) scl4300.1.1_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 4610131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4300.1.1_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4610131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2769462 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769462 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zznkm:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0