Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529025_s1
          See other OLFR1417 GE Assays ›
          Gene Symbol:
          Olfr1417
          Gene Name:
          olfactory receptor 1417
          Gene Aliases:
          MOR266-2
          Chromosome Location:
          Chr.19: 11828077 - 11829024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529025_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1417
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146936.1 NP_667147.1 1 895 83
          GenBank mRNA BC148189.1 - 1 895 40051854 83
          BC153816.1 - 1 895 40043087 83

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1417

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004821 NM_146936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004821 BC148189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004821 BC153816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004821 NM_146936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004821 BC148189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004821 BC153816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115139 NM_146936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115139 BC148189.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115139 BC153816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115139 BC150977.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115139 NM_001011524.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31958068 - -
          Whole Mouse Genome G4122A A_51_P115139 NM_146936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115139 BC148189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115139 BC153816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115139 BC150977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115139 NM_001011524.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14197432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14197420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31958064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14357446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14197418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14366510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14427321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31958043 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466267 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466266 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904128 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4920446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213096 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213096 - -
          mouse-6 v1.1 (Illumina) scl0258938.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258938.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213096 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0