Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529282_s1
          See other OLFR1233 GE Assays ›
          Gene Symbol:
          Olfr1233
          Gene Name:
          olfactory receptor 1233
          Gene Aliases:
          MOR238-1
          Chromosome Location:
          Chr.2: 89339312 - 89342391 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529282_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1233
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146972.1 NP_667183.1 1 828 104
          GenBank mRNA BC119149.1 - 1 876 8733898 104
          BC119151.1 - 1 876 8733900 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004054.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1233

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 BC119151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 NM_146972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 BC119151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 NM_146972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150648 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 BC119151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 NM_146972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 BC119149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 BC119151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 NM_146972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 BC119151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 NM_146972.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 BC119151.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 NM_146972.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150648 - -
          Whole Mouse Genome G4122A A_51_P466970 BC119151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466970 NM_146972.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466970 BC119149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466970 BC119151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466970 NM_146972.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P466970 BC119149.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124295 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21413485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01971444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21413512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01279064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484814 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484813 - -
          Affymetrix Mouse Clariom S Array PSR0200034039.mm.1 TC0200004054.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823589 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5220288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615495 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615495 - -
          mouse-6 v1.1 (Illumina) scl4281.1.1_216-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4281.1.1_216-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615495 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0