Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529359_s1
          See other OLFR1256 GE Assays ›
          Gene Symbol:
          Olfr1256
          Gene Name:
          olfactory receptor 1256
          Gene Aliases:
          MOR231-1
          Chromosome Location:
          Chr.2: 89835023 - 89835943 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529359_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1256
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146983.1 NP_667194.1 1 160 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004073.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1256

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179826 NM_146983.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131278 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179826 NM_146983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P199124 NM_146983.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131278 - -
          Whole Mouse Genome G4122A A_51_P199124 NM_146983.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121710 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21414903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01623306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01623316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01319943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02178217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01454448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02217587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01383877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01319942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01319944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21414947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484853 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484852 - -
          Affymetrix Mouse Clariom S Array PSR0200034109.mm.1 NM_146983.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034109.mm.1 NM_146983.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034109.mm.1 TC0200004073.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 749798 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2940195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940195 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254785 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254785 - -
          mouse-6 v1.1 (Illumina) scl4268.1.1_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4268.1.1_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254785 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254785 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline