Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529408_s1
          See other OLFR447 GE Assays ›
          Gene Symbol:
          Olfr447
          Gene Name:
          olfactory receptor 447
          Gene Aliases:
          MOR261-1
          Chromosome Location:
          Chr.6: 42911525 - 42912457 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529408_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr447
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146988.1 NP_667199.1 1 661 82
          GenBank mRNA BC104271.1 - 1 661 40040950 82
          BC104272.1 - 1 659 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000472.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr447

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025048 NM_146988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025048 BC104271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025048 BC104272.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117083 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025048 NM_146988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025048 BC104271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025048 BC104272.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P393288 NM_146988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P393288 BC104271.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P393288 BC104272.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400680 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117083 - -
          Whole Mouse Genome G4122A A_51_P393288 NM_146988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393288 BC104271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393288 BC104272.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106202 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05395015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05395041 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05395027 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05395032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05508755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05395037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05508762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05590993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05362489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05842697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05520325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05590995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537806 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537805 - -
          Affymetrix Mouse Clariom S Array PSR0600003742.mm.1 TC0600000472.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003742.mm.1 NM_146988.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003742.mm.1 NM_146988.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 565420 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2320398 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320398 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254919 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254919 - -
          mouse-6 v1.1 (Illumina) scl30118.1.1_232-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320398 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30118.1.1_232-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254919 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0