Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529417_s1
          See other OLFR1496 GE Assays ›
          Gene Symbol:
          Olfr1496
          Gene Name:
          olfactory receptor 1496
          Gene Aliases:
          MOR127-1
          Chromosome Location:
          Chr.19: 13780548 - 13781614 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529417_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1496
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146989.2 NP_667200.2 1 709 106
          GenBank mRNA BC137878.1 - 1 709 8860897 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000284.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1496

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051571 NM_146989.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051571 BC137878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2141073 NM_146989.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2141073 BC137878.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146893 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2141073 NM_146989.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2141073 BC137878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051571 NM_146989.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051571 BC137878.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205728 NM_146989.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205728 BC137878.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31965796 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146893 - -
          Whole Mouse Genome G4122A A_51_P205728 NM_146989.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205728 BC137878.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136020 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14270473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14145898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14145888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14145901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14145894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14145904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31965829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14145901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14145888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31965812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461833 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461832 - -
          Affymetrix Mouse Clariom S Array PSR1900003051.mm.1 NM_146989.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003051.mm.1 NM_146989.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003051.mm.1 TC1900000284.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 451869 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760072 - -
          Illumina mouse-6 v1.0 expression beadchip 6760072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696865 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696865 - -
          mouse-6 v1.1 (Illumina) scl6276.1.1_194-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6276.1.1_194-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696865 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0