Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529528_s1
          See other OLFR399 GE Assays ›
          Gene Symbol:
          Olfr399
          Gene Name:
          olfactory receptor 399
          Gene Aliases:
          MOR255-1
          Chromosome Location:
          Chr.11: 74053776 - 74054778 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529528_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr399
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147004.2 NP_667215.2 1 163 73
          GenBank mRNA BC120820.1 - 1 163 40129743 73
          BC120846.1 - 1 163 40129769 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr399

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111247 NM_147004.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111247 BC120820.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111247 BC120846.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101405 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111247 NM_147004.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111247 BC120820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111247 BC120846.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382647 NM_147004.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382647 BC120820.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P382647 BC120846.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101405 - -
          Whole Mouse Genome G4122A A_51_P382647 NM_147004.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382647 BC120820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P382647 BC120846.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128369 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101405 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09130207 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09130200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09650554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09130208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09202286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09650553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09650563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388301 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388300 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 722795 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2340594 - -
          Illumina mouse-6 v1.0 expression beadchip 2340594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755826 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755826 - -
          mouse-6 v1.1 (Illumina) scl39953.1_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39953.1_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755826 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755826 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0