Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529611_s1
          See other OLFR1038-PS GE Assays ›
          Gene Symbol:
          Olfr1038-ps
          Gene Name:
          olfactory receptor 1038, pseudogene
          Gene Aliases:
          MOR185-3, Olfr1038
          Chromosome Location:
          Chr.2: 86120224 - 86123037 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529611_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1038-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147013.2 NP_667224.1 2 147 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001268.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1038-ps

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974303 NM_147013.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641928 AK041693.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176437 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974303 NM_147013.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205155 NM_147013.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P205155 CB173938.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21400759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176437 - -
          Whole Mouse Genome G4122A A_51_P205155 NM_147013.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205155 CB173938.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107361 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02129949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01987821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01987833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01987837 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01757437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21400813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01606354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01589366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01606349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01589367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02155829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21400778 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473499 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473498 - -
          Affymetrix Mouse Clariom S Array PSR0200009995.mm.1 NM_147013.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009995.mm.1 NM_147013.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009995.mm.1 TC0200001268.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009996.mm.1 NM_147013.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009996.mm.1 NM_147013.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009996.mm.1 TC0200001268.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009997.mm.1 NM_147013.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009997.mm.1 NM_147013.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009997.mm.1 TC0200001268.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 298789 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000397 - -
          Illumina mouse-6 v1.0 expression beadchip 7000397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237162 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237162 - -
          mouse-6 v1.1 (Illumina) scl4927.1.1_160-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4927.1.1_160-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237162 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0