Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529621_s1
          See other OLFR1048 GE Assays ›
          Gene Symbol:
          Olfr1048
          Gene Name:
          olfactory receptor 1048
          Gene Aliases:
          MOR187-2
          Chromosome Location:
          Chr.2: 86235871 - 86236833 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529621_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1048
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147014.1 NP_667225.1 1 46 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003923.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1048

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966483 NM_147014.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405189 AK076394.1 genbank_mrna_mouse
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175103 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966483 NM_147014.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P367980 NM_147014.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175103 - -
          Whole Mouse Genome G4122A A_51_P367980 NM_147014.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111485 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21401358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01304238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21401423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01559141 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484576 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484575 - -
          Affymetrix Mouse Clariom S Array PSR0200033742.mm.1 NM_147014.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033742.mm.1 NM_147014.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033742.mm.1 TC0200003923.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 515300 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520537 - -
          Illumina mouse-6 v1.0 expression beadchip 520537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520537 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251051 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251051 - -
          mouse-6 v1.1 (Illumina) scl4343.1.1_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 520537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4343.1.1_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251051 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251051 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline