Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529698_s1
          See other OLFR378 GE Assays ›
          Gene Symbol:
          Olfr378
          Gene Name:
          olfactory receptor 378
          Gene Aliases:
          MOR135-2
          Chromosome Location:
          Chr.11: 73425037 - 73428477 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529698_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr378
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147024.2 NP_667235.1 2 706 121
          GenBank mRNA BC139813.1 - 1 575 40135620 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr378

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039791 NM_147024.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039791 BC139813.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168311 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039791 NM_147024.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039791 BC139813.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P274356 NM_147024.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P274356 BC139813.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27917062 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168311 - -
          Whole Mouse Genome G4122A A_51_P274356 NM_147024.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P274356 BC139813.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105169 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09441128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09441136 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09441138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09441133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09398547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09441147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09398548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27917030 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388268 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388267 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 565829 - -
          ABI Mouse Genome Survey Microarray 839958 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2741070 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2741070 - -
          mouse-6 v1.1 (Illumina) scl13068.1.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl13068.1.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2741070 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2741070 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl13068.1.1_24-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline