Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529721_s1
          See other OLFR1120 GE Assays ›
          Gene Symbol:
          Olfr1120
          Gene Name:
          olfactory receptor 1120
          Gene Aliases:
          MOR264-2
          Chromosome Location:
          Chr.2: 87354819 - 87358390 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529721_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147029.1 NP_667240.1 1 51 58
          GenBank mRNA BC119117.1 - 1 71 8733866 58
          BC119143.1 - 1 71 8733892 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001287.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1120

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976460 NM_147029.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976460 BC119117.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976460 BC119143.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154484 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976460 NM_147029.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976460 BC119117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976460 BC119143.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109357 NM_147029.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109357 BC119117.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109357 BC119143.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405648 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154484 - -
          Whole Mouse Genome G4122A A_51_P109357 NM_147029.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109357 BC119117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109357 BC119143.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118248 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01381999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01382001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01381980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01382000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01381992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01382008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01381998 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01381989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01251302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01186121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01330548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01381989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473529 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473528 - -
          Affymetrix Mouse Clariom S Array PSR0200010031.mm.1 NM_147029.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010031.mm.1 NM_147029.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010031.mm.1 TC0200001287.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777143 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1170427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2654348 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2654348 - -
          mouse-6 v1.1 (Illumina) scl4917.1.1_188-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4917.1.1_188-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2654348 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2654348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline