Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529781_s1
          See other OLFR711 GE Assays ›
          Gene Symbol:
          Olfr711
          Gene Name:
          olfactory receptor 711
          Gene Aliases:
          MOR103-4
          Chromosome Location:
          Chr.7: 106971368 - 106975451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529781_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr711
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147035.2 NP_667246.2 3 846 88
          GenBank mRNA BC132382.1 - 1 40130659 88
          BC138433.1 - 1 8861453 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr711

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404281 BC132382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404281 BC132382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165696 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404281 BC132382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404281 BC138433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404281 BC132382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P404281 BC132382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P404281 BC132382.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165696 - -
          Whole Mouse Genome G4122A A_52_P404281 BC132382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404281 BC138433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404281 BC132382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404281 BC138433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124130 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06422733 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06658488 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06372829 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06658495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06415890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06299894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06229882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06138744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397424 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566621 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566620 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 336167 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5860068 - -
          Illumina mouse-6 v1.0 expression beadchip 5860068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769318 - -
          mouse-6 v1.1 (Illumina) scl8840.1.1_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8840.1.1_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2963526 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2963526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0