Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529864_s1
          See other OLFR600 GE Assays ›
          Gene Symbol:
          Olfr600
          Gene Name:
          olfactory receptor 600
          Gene Aliases:
          MOR39-1
          Chromosome Location:
          Chr.7: 103345982 - 103346926 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529864_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr600
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147046.2 NP_667257.2 1 245 77
          GenBank mRNA BC130247.1 - 1 249 40142365 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003884.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr600

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163671 NM_147046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163671 NM_147046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216161 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140115 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163671 NM_147046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163671 BC130247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163671 NM_147046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P163671 NM_147046.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P163671 NM_147046.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25382970 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140115 - -
          Whole Mouse Genome G4122A A_51_P163671 NM_147046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163671 BC130247.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163671 NM_147046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P163671 BC130247.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116039 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05892995 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05892994 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05893009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06610939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05893006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06376657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05892999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382960 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566208 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566207 - -
          Affymetrix Mouse Clariom S Array PSR0700032695.mm.1 NM_147046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032695.mm.1 NM_147046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032695.mm.1 TC0700003884.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 875697 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2825410 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2825410 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0