Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529946_s1
          See other OLFR649 GE Assays ›
          Gene Symbol:
          Olfr649
          Gene Name:
          olfactory receptor 649
          Gene Aliases:
          MOR31-7
          Chromosome Location:
          Chr.7: 104189267 - 104190205 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529946_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr649
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147055.1 NP_667266.1 1 50 59
          GenBank mRNA BC104080.2 - 1 43 59
          BC104081.2 - 1 43 59
          BC119409.2 - 1 43 59
          BC119410.2 - 1 43 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003934.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr649

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180685 NM_147055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180685 BC104080.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180685 BC104081.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180685 BC119409.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180685 BC119410.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123960 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180685 NM_147055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180685 BC104080.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180685 BC104081.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180685 BC119409.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180685 BC119410.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273234 NM_147055.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273234 BC104080.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273234 BC104081.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273234 BC119409.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273234 BC119410.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387067 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123960 - -
          Whole Mouse Genome G4122A A_51_P273234 NM_147055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273234 BC104080.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273234 BC104081.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273234 BC119409.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273234 BC119410.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125415 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125197 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05885614 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06158415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06158414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06172466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06172460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06114142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387063 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566325 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566324 - -
          Affymetrix Mouse Clariom S Array PSR0700032866.mm.1 NM_147055.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032866.mm.1 NM_147055.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032866.mm.1 TC0700003934.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316006 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619810 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619810 - -
          mouse-6 v1.1 (Illumina) scl8860.1.1_248-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8860.1.1_248-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619810 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline