Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529982_s1
          See other OLFR667 GE Assays ›
          Gene Symbol:
          Olfr667
          Gene Name:
          olfactory receptor 667
          Gene Aliases:
          MOR34-11, MOR34-2
          Chromosome Location:
          Chr.7: 104916314 - 104917294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529982_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr667
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147060.2 NP_667271.2 1 642 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003958.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr667

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2084092 NM_147060.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698465 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166749 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2084092 NM_147060.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P383063 NM_147060.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25389904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166749 - -
          Whole Mouse Genome G4122A A_51_P383063 NM_147060.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06386725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25389873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06469285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05964236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05964240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06083249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06064425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25389822 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566402 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566400 - -
          Affymetrix Mouse Clariom S Array PSR0700033038.mm.1 NM_147060.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033038.mm.1 NM_147060.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033038.mm.1 TC0700003958.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 567247 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690040 - -
          Illumina mouse-6 v1.0 expression beadchip 2690040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710932 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710932 - -
          mouse-6 v1.1 (Illumina) scl0259062.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259062.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213880 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213880 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0