Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529996_s1
          See other OLFR124 GE Assays ›
          Gene Symbol:
          Olfr124
          Gene Name:
          olfactory receptor 124
          Gene Aliases:
          A3, MOR256-3
          Chromosome Location:
          Chr.17: 37805069 - 37806190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529996_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr124
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147062.2 NP_667273.1 1 228 110
          GenBank mRNA BC119256.1 - 1 228 8734005 110
          BC119258.1 - 1 228 8734007 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000785.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr124

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980661 NM_147062.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980661 BC119256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980661 BC119258.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142399 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980661 NM_147062.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980661 BC119256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980661 BC119258.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228936 NM_147062.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228936 BC119256.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228936 BC119258.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31170000 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142399 - -
          Whole Mouse Genome G4122A A_51_P228936 NM_147062.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228936 BC119256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228936 BC119258.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135648 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501339 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13501335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31170047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13220980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13378941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13205554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13156586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31170040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445160 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445158 - -
          Affymetrix Mouse Clariom S Array PSR1700007747.mm.1 NM_147062.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007747.mm.1 NM_147062.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007747.mm.1 TC1700000785.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538981 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6400195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400195 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774404 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774404 - -
          mouse-6 v1.1 (Illumina) scl8512.1.1_125-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8512.1.1_125-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2822080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774404 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2822080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline