Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530041_s1
          See other OLFR829 GE Assays ›
          Gene Symbol:
          Olfr829
          Gene Name:
          olfactory receptor 829
          Gene Aliases:
          MOR147-1
          Chromosome Location:
          Chr.9: 18855062 - 18857592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530041_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr829
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147067.1 NP_667278.1 1 751 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr829

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027047 NM_147067.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729551 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111767 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027047 NM_147067.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127094 NM_147067.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338584 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111767 - -
          Whole Mouse Genome G4122A A_51_P127094 NM_147067.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121300 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111767 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08073451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08073452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07528354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07904604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08187241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08073454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07904603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338595 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583377 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583376 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 882088 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190253 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688469 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688469 - -
          mouse-6 v1.1 (Illumina) scl37256.1.28_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190253 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37256.1.28_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190253 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2688469 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2688469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline