Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530054_s1
          See other OLFR686 GE Assays ›
          Gene Symbol:
          Olfr686
          Gene Name:
          olfactory receptor 686
          Gene Aliases:
          MOR35-1
          Chromosome Location:
          Chr.7: 105203388 - 105204341 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530054_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr686
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147069.1 NP_667280.1 1 453 76
          GenBank mRNA BC132004.1 - 1 488 40130281 76
          BC132006.1 - 1 488 40130283 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003972.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr686

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996978 BC132004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996978 BC132006.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996978 NM_147069.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121163 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996978 BC132004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996978 BC132006.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996978 NM_147069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301236 BC132004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301236 BC132006.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P301236 NM_147069.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25391243 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121163 - -
          Whole Mouse Genome G4122A A_51_P301236 BC132004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301236 BC132006.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301236 NM_147069.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104415 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06182960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06182963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06182959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06182969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25391248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06182973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06281995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06314228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25391289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566433 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566432 - -
          Affymetrix Mouse Clariom S Array PSR0700033061.mm.1 NM_147069.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033061.mm.1 NM_147069.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033061.mm.1 TC0700003972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033062.mm.1 TC0700003972.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 615271 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000400 - -
          Illumina mouse-6 v1.0 expression beadchip 7000400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2654109 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2654109 - -
          mouse-6 v1.1 (Illumina) scl8849.1.1_76-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8849.1.1_76-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2654109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2654109 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline