Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530088_s1
          See other OLFR656 GE Assays ›
          Gene Symbol:
          Olfr656
          Gene Name:
          olfactory receptor 656
          Gene Aliases:
          MOR27-1
          Chromosome Location:
          Chr.7: 104617657 - 104618646 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530088_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr656
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147075.1 NP_667286.1 1 941 62
          GenBank mRNA BC110472.1 - 1 941 40044290 62
          BC110473.2 - 1 940 40044291 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001527.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr656

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134471 NM_147075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134471 BC110472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2134471 BC110473.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157142 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134471 NM_147075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134471 BC110472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2134471 BC110473.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P351828 NM_147075.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P351828 BC110472.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P351828 BC110473.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25388843 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157142 - -
          Whole Mouse Genome G4122A A_51_P351828 NM_147075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351828 BC110472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351828 BC110473.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138194 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06438069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05992284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06509660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388822 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555880 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555879 - -
          Affymetrix Mouse Clariom S Array PSR0700012900.mm.1 TC0700001527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012900.mm.1 NM_147075.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012900.mm.1 NM_147075.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 387735 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4730301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218528 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218528 - -
          mouse-6 v1.1 (Illumina) scl9385.1.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9385.1.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218528 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218528 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline