Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530097_s1
          See other OLFR619 GE Assays ›
          Gene Symbol:
          Olfr619
          Gene Name:
          olfactory receptor 619
          Gene Aliases:
          MOR31-5
          Chromosome Location:
          Chr.7: 103603589 - 103604712 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530097_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr619
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147076.2 NP_667287.2 1 915 62
          GenBank mRNA BC141053.1 - 1 915 9055665 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001502.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr619

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986681 NM_147076.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986681 BC141053.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114864 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986681 NM_147076.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986681 BC141053.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P131827 NM_147076.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P131827 BC141053.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114864 - -
          Whole Mouse Genome G4122A A_51_P131827 NM_147076.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P131827 BC141053.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104638 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25384118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06291202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25384118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555806 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555805 - -
          Affymetrix Mouse Clariom S Array PSR0700012765.mm.1 TC0700001502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012765.mm.1 NM_147076.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012765.mm.1 NM_147076.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012764.mm.1 TC0700001502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012764.mm.1 NM_147076.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012764.mm.1 NM_147076.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 330387 - -
          ABI Mouse Genome Survey Microarray 358323 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714128 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714128 - -
          mouse-6 v1.1 (Illumina) scl0259080.1_63-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259080.1_63-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714128 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline