Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530132_s1
          See other OLFR610 GE Assays ›
          Gene Symbol:
          Olfr610
          Gene Name:
          olfactory receptor 610
          Gene Aliases:
          MOR9-2
          Chromosome Location:
          Chr.7: 103505997 - 103506944 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530132_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr610
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147081.2 NP_667292.2 1 450 102
          GenBank mRNA BC113199.1 - 1 449 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003893.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr610

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P538412 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P538412 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P538412 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P319114 NM_147081.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P319114 BC113199.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383577 - -
          Whole Mouse Genome G4122A A_51_P319114 NM_147081.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P319114 BC113199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P538412 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110982 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06525471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06278936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06350457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566220 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566219 - -
          Affymetrix Mouse Clariom S Array PSR0700032718.mm.1 NM_147081.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032718.mm.1 NM_147081.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032718.mm.1 TC0700003893.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 366978 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160741 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160741 - -
          mouse-6 v1.1 (Illumina) GI_22128924-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160741 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160741 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline