Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530166_s1
          See other OLFR639 GE Assays ›
          Gene Symbol:
          Olfr639
          Gene Name:
          olfactory receptor 639
          Gene Aliases:
          MOR12-1
          Chromosome Location:
          Chr.7: 104011750 - 104012700 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530166_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr639
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147084.1 NP_667295.1 1 901 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr639

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960876 NM_147084.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582367 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960876 NM_147084.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P63643 NM_147084.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386191 - -
          Whole Mouse Genome G4122A A_52_P63643 NM_147084.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566293 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566292 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 798432 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850528 - -
          Illumina mouse-6 v1.0 expression beadchip 4850528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218318 - -
          mouse-6 v1.1 (Illumina) scl8865.1.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8865.1.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2940110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2940110 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline