Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530203_s1
          See other OLFR569 GE Assays ›
          Gene Symbol:
          Olfr569
          Gene Name:
          olfactory receptor 569
          Gene Aliases:
          MOR30-1
          Chromosome Location:
          Chr.7: 102887207 - 102888151 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530203_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr569
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147088.1 NP_667299.1 1 907 83
          GenBank mRNA BC126864.1 - 1 907 40042577 83
          BC126865.1 - 1 907 40042578 83
          BC127048.1 - 1 907 40056696 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr569

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158906 BC126865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158906 BC127048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158906 NM_147088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158906 BC126864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050822 BC126865.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050822 BC127048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050822 NM_147088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050822 BC126864.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114680 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050822 BC126865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050822 BC127048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050822 NM_147088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050822 BC126864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158906 BC126865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158906 BC127048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158906 NM_147088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158906 BC126864.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347206 BC126865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347206 BC127048.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347206 NM_147088.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P347206 BC126864.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25381035 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114680 - -
          Whole Mouse Genome G4122A A_51_P347206 BC126865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347206 BC127048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347206 NM_147088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347206 BC126864.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127079 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171560 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06171567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06171547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381028 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566185 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566184 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632273 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170047 - -
          Illumina mouse-6 v1.0 expression beadchip 1170047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231609 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231609 - -
          mouse-6 v1.1 (Illumina) scl8890.1.1_256-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8890.1.1_256-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231609 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline