Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530228_s1
          See other OLFR568 GE Assays ›
          Gene Symbol:
          Olfr568
          Gene Name:
          olfactory receptor 568
          Gene Aliases:
          MOR14-11, MOR14-3
          Chromosome Location:
          Chr.7: 102877122 - 102878063 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530228_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr568
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147091.2 NP_667302.2 1 634 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001469.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr568

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974923 NM_147091.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110880 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974923 NM_147091.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P285418 NM_147091.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110880 - -
          Whole Mouse Genome G4122A A_51_P285418 NM_147091.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127492 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108505 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06040845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06099463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06186967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380994 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555747 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555746 - -
          Affymetrix Mouse Clariom S Array PSR0700012700.mm.1 TC0700001469.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012700.mm.1 NM_147091.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012700.mm.1 NM_147091.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 307892 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7050494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646854 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646854 - -
          mouse-6 v1.1 (Illumina) scl9407.1.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9407.1.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646854 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646854 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0