Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530232_s1
          See other OLFR561 GE Assays ›
          Gene Symbol:
          Olfr561
          Gene Name:
          olfactory receptor 561
          Gene Aliases:
          MOR14-2
          Chromosome Location:
          Chr.7: 102774526 - 102775470 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530232_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr561
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147092.1 NP_667303.1 1 54 84
          GenBank mRNA BC120569.1 - 1 85 40129492 84
          BC120571.1 - 1 85 40129494 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001467.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr561

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993228 BC120569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993228 BC120571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993228 NM_147092.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101438 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993228 BC120569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993228 BC120571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993228 NM_147092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358507 BC120569.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358507 BC120571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P358507 NM_147092.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101438 - -
          Whole Mouse Genome G4122A A_51_P358507 BC120569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358507 BC120571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358507 NM_147092.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129067 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06156706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06156711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06198842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06614148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06553769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25380590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555743 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555741 - -
          Affymetrix Mouse Clariom S Array PSR0700012696.mm.1 TC0700001467.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012697.mm.1 NM_147092.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012697.mm.1 NM_147092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012697.mm.1 TC0700001467.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358501 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2714080 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2714080 - -
          mouse-6 v1.1 (Illumina) scl9410.1.1_296-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9410.1.1_296-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2714080 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline