Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530319_s1
          See other OLFR555 GE Assays ›
          Gene Symbol:
          Olfr555
          Gene Name:
          olfactory receptor 555
          Gene Aliases:
          MOR10-1
          Chromosome Location:
          Chr.7: 102658823 - 102659770 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530319_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr555
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147103.2 NP_667314.2 1 349 116
          GenBank mRNA BC106981.1 - 1 349 40043127 116
          BC106982.1 - 1 349 40043129 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001463.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr555

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041554 NM_147103.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041554 BC106981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041554 BC106982.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103119 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041554 NM_147103.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041554 BC106981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041554 BC106982.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P101950 NM_147103.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P101950 BC106981.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P101950 BC106982.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25379928 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103119 - -
          Whole Mouse Genome G4122A A_51_P101950 NM_147103.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P101950 BC106981.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P101950 BC106982.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140714 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138997 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25380001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06106298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06106296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379921 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555730 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555729 - -
          Affymetrix Mouse Clariom S Array PSR0700012688.mm.1 TC0700001463.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012688.mm.1 NM_147103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012688.mm.1 NM_147103.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 551349 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4920154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2767446 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2767446 - -
          mouse-6 v1.1 (Illumina) scl9412.1.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9412.1.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2767446 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2767446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline