Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530333_s1
          See other OLFR550 GE Assays ›
          Gene Symbol:
          Olfr550
          Gene Name:
          olfactory receptor 550
          Gene Aliases:
          MOR16-1
          Chromosome Location:
          Chr.7: 102571431 - 102579662 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530333_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr550
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147104.2 NP_667315.2 1 879 62
          GenBank mRNA BC116960.1 - 1 879 40126279 62
          BC116964.1 - 1 879 40126283 62

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr550

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111100 BC116960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111100 BC116964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111100 NM_147104.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147158 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111100 BC116960.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111100 BC116964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111100 NM_147104.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279827 BC116960.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279827 BC116964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279827 NM_147104.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147158 - -
          Whole Mouse Genome G4122A A_51_P279827 BC116960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279827 BC116964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279827 NM_147104.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106520 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25379637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06650123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05932352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06236637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06069889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06499001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06119805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06082750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06119806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06284441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25379605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555723 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555722 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 472189 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940600 - -
          Illumina mouse-6 v1.0 expression beadchip 6940600 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2684838 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2684838 - -
          mouse-6 v1.1 (Illumina) scl9415.1.1_190-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940600 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9415.1.1_190-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2684838 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2684838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline