Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530344_s1
          See other OLFR978 GE Assays ›
          Gene Symbol:
          Olfr978
          Gene Name:
          olfactory receptor 978
          Gene Aliases:
          MOR223-3
          Chromosome Location:
          Chr.9: 39993812 - 39994747 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530344_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr978
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147105.2 NP_667316.2 1 871 93
          GenBank mRNA BC114537.1 - 1 870 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000443.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr978

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134297 NM_147105.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134297 BC114537.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128615 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134297 NM_147105.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134297 BC114537.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P196213 NM_147105.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P196213 BC114537.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440847 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128615 - -
          Whole Mouse Genome G4122A A_51_P196213 NM_147105.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196213 BC114537.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134297 NM_147105.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134297 BC114537.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135360 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128615 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07812369 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07677364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07812367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07677375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07812370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07622452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07677364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440829 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584521 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584520 - -
          Affymetrix Mouse Clariom S Array PSR0900002919.mm.1 NM_147105.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002919.mm.1 NM_147105.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002919.mm.1 TC0900000443.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894559 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3440520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761233 - -
          mouse-6 v1.1 (Illumina) scl7555.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7555.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2761233 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2761233 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0