Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530363_s1
          See other OLFR974 GE Assays ›
          Gene Symbol:
          Olfr974
          Gene Name:
          olfactory receptor 974
          Gene Aliases:
          MOR171-1
          Chromosome Location:
          Chr.9: 39942262 - 39943194 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530363_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr974
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147107.1 NP_667318.1 1 855 106
          GenBank mRNA BC131914.1 - 1 902 40130191 106
          BC132593.1 - 1 902 40130870 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000442.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr974

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 NM_147107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 BC131914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 NM_147107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 BC131914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161559 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 NM_147107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 BC131914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 BC132593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 NM_147107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 BC131914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 NM_147107.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 BC131914.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 NM_147107.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 BC131914.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26440561 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161559 - -
          Whole Mouse Genome G4122A A_52_P672636 NM_147107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P672636 BC131914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P672636 BC132593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P672636 NM_147107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P672636 BC131914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P672636 BC132593.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133164 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07482169 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07482173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07482156 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07482175 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07482153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26440583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07798382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07798402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26440609 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584519 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584518 - -
          Affymetrix Mouse Clariom S Array PSR0900002917.mm.1 TC0900000442.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 539784 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219329 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219329 - -
          mouse-6 v1.1 (Illumina) scl37073.1.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37073.1.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219329 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219329 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0