Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530385_s1
          See other OLFR570 GE Assays ›
          Gene Symbol:
          Olfr570
          Gene Name:
          olfactory receptor 570
          Gene Aliases:
          MOR8-3
          Chromosome Location:
          Chr.7: 102813310 - 102901380 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00530385_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr570
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147110.1 NP_667321.1 1 50 88
          GenBank mRNA BC116966.1 - 1 77 40126285 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001470.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr570

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981589 NM_147110.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981589 BC116966.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677727 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126779 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981589 NM_147110.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981589 BC116966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352864 NM_147110.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352864 BC116966.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126779 - -
          Whole Mouse Genome G4122A A_51_P352864 NM_147110.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352864 BC116966.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114454 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25381131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06275489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06384092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06585505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25381095 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555750 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555749 - -
          Affymetrix Mouse Clariom S Array PSR0700012701.mm.1 TC0700001470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012702.mm.1 TC0700001470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012702.mm.1 NM_147110.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012702.mm.1 NM_147110.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 600126 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4120025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636578 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636578 - -
          mouse-6 v1.1 (Illumina) scl9406.1.1_211-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9406.1.1_211-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636578 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636578 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0