Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00545980_s1
          See other SEPHS2 GE Assays ›
          Gene Symbol:
          Sephs2
          Gene Name:
          selenophosphate synthetase 2
          Gene Aliases:
          Sps2, Ysg3
          Chromosome Location:
          Chr.7: 127271879 - 127274059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00545980_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sephs2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009266.3 NP_033292.2 1 758 67
          GenBank mRNA AK077573.1 - 1 759 67
          AK149516.1 - 1 755 67
          AK168722.1 - 1 759 67
          AK168965.1 - 1 759 67
          BC003760.1 - 1 745 67
          BC016508.1 - 1 750 4911695 67
          BC028966.1 - 1 735 5374184 67
          BC043334.1 - 1 739 5100207 67
          U43285.1 - 1 755 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004321.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sephs2

          Panther Classification:

          Molecular Function -

          transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          selenium compound metabolic process
          selenocysteine biosynthetic process
          phosphorylation
          nucleotide binding
          catalytic activity
          selenide, water dikinase activity
          ATP binding
          kinase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504898 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504900 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504894 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079324 BC043334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079324 U43285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079324 BC016508.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079324 BC028966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079324 AK149516.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596924 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130102 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079324 BC043334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079324 U43285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079324 BC016508.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079324 BC028966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079324 AK149516.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130102 - -
          Whole Mouse Genome G4122A A_51_P418765 BC043334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418765 U43285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418765 BC016508.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418765 BC028966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418765 AK149516.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132839 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06066795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05907328 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05907330 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06066797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504900 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06011277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05907330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06320804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06474371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05907324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06011275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05907328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06612374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06457546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06223069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06066797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05945318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05907336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25504877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06640182 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568224 - -
          Affymetrix HT MG-430 PM Array Plate 1418325_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449040_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568221 - -
          Mouse Genome 430 2.0 Array 1435602_at - -
          Mouse Genome 430 2.0 Array 1418325_at - -
          Mouse Genome 430 2.0 Array 1449040_PM_a_at - -
          Mouse Genome 430 2.0 Array 1435602_PM_at - -
          Mouse Genome 430 2.0 Array 1449040_a_at - -
          Mouse Genome 430 2.0 Array 1418325_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20768_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20768_at - -
          Affymetrix Mouse Clariom S Array PSR0700036293.mm.1 NM_009266.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036293.mm.1 NM_009266.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036293.mm.1 TC0700004321.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036294.mm.1 NM_009266.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036294.mm.1 NM_009266.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036294.mm.1 TC0700004321.mm.1 MTA TC
          Mouse Expression Set 430 1435602_PM_at - -
          Mouse Expression Set 430 1449040_a_at - -
          Mouse Expression Set 430 1418325_at - -
          Mouse Expression Set 430 1449040_PM_a_at - -
          Mouse Expression Set 430 1435602_at - -
          Mouse Expression Set 430 1418325_PM_at - -
          Murine Genome U74v2 Set 93557_PM_at - -
          Murine Genome U74v2 Set 93557_at - -
          Mouse Genome 430A 2.0 Array 1435602_at - -
          Mouse Genome 430A 2.0 Array 1418325_PM_at - -
          Mouse Genome 430A 2.0 Array 1418325_at - -
          Mouse Genome 430A 2.0 Array 1449040_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1449040_a_at - -
          Mouse Genome 430A 2.0 Array 1435602_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435602_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418325_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435602_at - -
          Affymetrix HT Mouse Genome 430A Array 1418325_at - -
          Affymetrix HT Mouse Genome 430A Array 1449040_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449040_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 860165 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770446 - -
          Illumina mouse-6 v1.0 expression beadchip 6770446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217590 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217590 - -
          mouse-6 v1.1 (Illumina) scl30651.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30651.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217590 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217590 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline