Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546511_m1
          See other SDHD GE Assays ›
          Gene Symbol:
          Sdhd
          Gene Name:
          succinate dehydrogenase complex, subunit D, integral membrane protein
          Gene Aliases:
          3110001M13Rik, AVLL5809, C78570, PRO19626
          Chromosome Location:
          Chr.9: 50596340 - 50603849 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546511_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdhd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025848.3 NP_080124.1 1 - 2 117 70
          GenBank mRNA AK013962.2 - 1 - 2 85 70
          AK135970.1 - 1 - 2 77 70
          BC145731.1 - 1 - 2 77 40131034 70
          BC145733.1 - 1 - 2 77 40131036 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002271.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdhd

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          transport
          regulation of catecholamine secretion
          oxidation-reduction process
          cellular response to hypoxia
          succinate dehydrogenase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26504536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26504535 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK013962.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK135970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK013503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 BF662071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK013962.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK135970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 AK013503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 BF662071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6475 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131083 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK013962.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK135970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK013503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 BF662071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 NM_025848.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK013962.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK135970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 AK013503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 BF662071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK013962.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK135970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK013503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 BF662071.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK013962.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK135970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 AK013503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 BF662071.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26504509 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06785286 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131083 - -
          Whole Mouse Genome G4122A A_51_P260871 AK013962.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 AK135970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 AK013503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 BF662071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 NM_025848.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 AK013962.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 AK135970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 AK013503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 BF662071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260871 NM_025848.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123508 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07683760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08035969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07683769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08035978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26504536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26504535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26504512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08078735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07689181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07732769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07683769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07532637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08035978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07910969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26504497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593358 - -
          Affymetrix HT MG-430 PM Array Plate 1437489_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1428235_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593356 - -
          Mouse Genome 430 2.0 Array 1428235_at - -
          Mouse Genome 430 2.0 Array 1428235_PM_at - -
          Mouse Genome 430 2.0 Array 1437489_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437489_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66925_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66925_at - -
          Affymetrix Mouse Clariom S Array PSR0900018886.mm.1 NM_025848.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018886.mm.1 NM_025848.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018886.mm.1 TC0900002271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018884.mm.1 NM_025848.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018884.mm.1 NM_025848.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018884.mm.1 TC0900002271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018885.mm.1 NM_025848.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018885.mm.1 NM_025848.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018885.mm.1 TC0900002271.mm.1 MTA TC
          Mouse Expression Set 430 1437489_PM_x_at - -
          Mouse Expression Set 430 1428235_at - -
          Mouse Expression Set 430 1428235_PM_at - -
          Mouse Expression Set 430 1437489_x_at - -
          Murine Genome U74v2 Set 92574_PM_at - -
          Murine Genome U74v2 Set 92574_at - -
          Murine Genome U74v2 Set 169663_PM_at - -
          Murine Genome U74v2 Set 169663_at - -
          Mouse Genome 430A 2.0 Array 1437489_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437489_x_at - -
          Mouse Genome 430A 2.0 Array 1428235_PM_at - -
          Mouse Genome 430A 2.0 Array 1428235_at - -
          Affymetrix HT Mouse Genome 430A Array 1437489_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437489_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1428235_at - -
          Affymetrix HT Mouse Genome 430A Array 1428235_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 557782 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2570128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239143 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239143 - -
          mouse-6 v1.1 (Illumina) scl35879.4_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35879.4_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239143 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239143 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35879.4_19-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline